Learn More
Perkin Elmer US LLC 5-Position Priority Samples Rack
Catalog No. 502112656
Encompass_Preferred
This rack features inserts for 8 mL tube (N8145125), 15 mL tube (N0777471), 50 mL tube (N8145124), 125 mL bottle (N0777820) and 250 mL bottle (N0777818). For PerkinElmer prepFAST only.
Catalog No. 50-211-2656 Supplier Perkin Elmer US LLC Supplier No. N8145317
This rack features inserts for 8 mL tube (N8145125), 15 mL tube (N0777471), 50 mL tube (N8145124), 125 mL bottle (N0777820) and 250 mL bottle (N0777818). For PerkinElmer prepFAST only.