Learn More
Medchemexpress LLC (5R)-3-(3-fluoro-4-morpholin-4-ylphenyl)-5-(hydroxymethyl)-1,3-oxazolidin-2-one | 168828-82-8 | MFCD06658241 | 99.7% | 296.29 g/mol | C14H17FN2O4 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01970010G
Encompass_Preferred
PNU-100440 is a small-molecule metabolite of the antibiotic linezolid (CAS 168828-82-8). It is provided for laboratory research use as a solid or as a DMSO stock solution and is intended for in vitro studies, metabolite analysis, and analytical reference purposes.
  • Chemical formula C14H17FN2O4 and molecular weight 296.29 g/mol.
  • High chemical purity (99.67%).
  • Available as solid and 10 mM solution in DMSO.
  • Offered in multiple pack sizes including gram-scale quantities.
  • Suitable for metabolite identification and analytical reference standards.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-99549