Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone, 98%

Catalog No. AAH6082103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6082103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6082103 Supplier Thermo Scientific Chemicals Supplier No. H6082103
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 100516-54-9 | C24H21NO4 | 387.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100516-54-9
Molecular Formula C24H21NO4
Molecular Weight (g/mol) 387.44
MDL Number MFCD00074958
InChI Key HECRUWTZAMPQOS-UHFFFAOYNA-N
Synonym 2r,3s-benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate, benzyl 2r,3s---6-oxo-2,3-diphenyl-4-morpholinecarboxylate, benzyl 2r,3s-6-oxo-2,3-diphenylmorpholine-4-carboxylate, 2r,3s-n-cbz-6-oxo-2,3-diphenylmorpholine, 2r,3s---n-z-6-oxo-2,3-diphenylmorpholine, benzyl 2r,3s-6-oxo-2,3-diphenyl-4-morpholinecarboxylate, pubchem18045, 2r,3s-6-oxo-2,3-diphenyl-4-morpholinecarboxylate
PubChem CID 981238
IUPAC Name benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate
SMILES O=C(OCC1=CC=CC=C1)N1CC(=O)OC(C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 205°C to 207°C
Quantity 1 g
Formula Weight 387.43
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.