Learn More
Medchemexpress LLC (5S,6R)-Methyl 5,6,7-trihydroxyheptanoate | 78606-80-1 | MFCD00211214 | >98.0% | 192.21 g/mol | C8H16O5 | 1 ML
SDP
Catalog No. 5000214925
Encompass_Preferred
BML-111 is a lipoxin A4 analog and a potent lipoxin A4 receptor (FPR2/ALX) agonist used in research to modulate inflammation, angiogenesis, and immune responses. It is supplied in research-grade formats with high purity and is suitable for in vitro pharmacology and cell signaling studies.
  • High purity suitable for research use.
  • Available as a 10 mM solution in DMSO and as solid forms.
  • Molecular weight 192.21 g/mol; molecular formula C8H16O5.
  • Used to study lipoxin signaling, inflammation, and tumor models.
  • Provided in small laboratory quantities for easy handling and dosing.
Catalog No. 50-002-14925 Supplier Medchemexpress LLC Supplier No. HY10045010MM 1ML
BML-111 is a lipoxin A4 analog and a potent lipoxin A4 receptor (FPR2/ALX) agonist used in research to modulate inflammation, angiogenesis, and immune responses. It is supplied in research-grade formats with high purity and is suitable for in vitro pharmacology and cell signaling studies.
  • High purity suitable for research use.
  • Available as a 10 mM solution in DMSO and as solid forms.
  • Molecular weight 192.21 g/mol; molecular formula C8H16O5.
  • Used to study lipoxin signaling, inflammation, and tumor models.
  • Provided in small laboratory quantities for easy handling and dosing.