Learn More
Enzo Life Sciences 5(S)-Hydroxyeicosa-6E,8Z,11Z,14Z-tetraenoic acid (HETE) (50µg). CAS: 70608-72-9
SDP
Supplier:  Enzo Life Sciences BMLH0050050
Encompass_Preferred
Biosynthesis via 5-lipoxygenase. Chemotactic and chemokinetic for PMNL. Induces degranulation, and calcium flux. Alternative name: 5(S)-HETE. Formulation: Liquid. Solution in ethanol. Purity: ≥98%. Long Term Storage: -80°C.
Catalog No. 50-201-1016