Learn More
Medchemexpress LLC 5s rRNA modificator (FAI) | 1415238-77-5 | 95.6% | 176.17 | 1 ML
SDP
Catalog No. 5000387766
Encompass_Preferred
5S rRNA modificator is a suitable electrophile for 2'-hydroxyl acylation on structured RNA molecules, yielding accurate structural information comparable to that obtained with existing probes. It is used for 5S rRNA RNA modification.
  • Suitable electrophile for 2'-hydroxyl acylation
  • Yields accurate structural information
  • Comparable to existing probes
  • Used for RNA modification
Catalog No. 50-003-87766 Supplier Medchemexpress LLC Supplier No. HY1840810MM/1ML
5S rRNA modificator is a suitable electrophile for 2'-hydroxyl acylation on structured RNA molecules, yielding accurate structural information comparable to that obtained with existing probes. It is used for 5S rRNA RNA modification.
  • Suitable electrophile for 2'-hydroxyl acylation
  • Yields accurate structural information
  • Comparable to existing probes
  • Used for RNA modification