Learn More
Medchemexpress LLC 5S rRNA modificator (FAI) | 1415238-77-5 | 95.61% | 176.17 | 1 MG
SDP
Catalog No. 5000387767
Encompass_Preferred
5S rRNA modificator is an electrophile suitable for 2'-hydroxyl acylation on structured RNA molecules. It yields accurate structural information comparable to that obtained with existing probes, and is used in 5S rRNA RNA modification research. For research use only.
  • Suitable for 2'-hydroxyl acylation on structured RNA molecules
  • Provides accurate structural information
  • Used in RNA modification research
Catalog No. 50-003-87767 Supplier Medchemexpress LLC Supplier No. HY184081MG
5S rRNA modificator is an electrophile suitable for 2'-hydroxyl acylation on structured RNA molecules. It yields accurate structural information comparable to that obtained with existing probes, and is used in 5S rRNA RNA modification research. For research use only.
  • Suitable for 2'-hydroxyl acylation on structured RNA molecules
  • Provides accurate structural information
  • Used in RNA modification research