Learn More
Aat Bioquest 5-SFX [5-FAM-X, SE] [CAS 148356-00-7]
SDP
Catalog No. 502979167
Encompass_Preferred
5-FAM-X, SE contains the aminohexanoyl spacer (known as 'X') between the FAM fluorophore and the succinimidyl ester.
Catalog No. 50-297-9167 Supplier Aat Bioquest Supplier No. 119
5-FAM-X, SE contains the aminohexanoyl spacer (known as 'X') between the FAM fluorophore and the succinimidyl ester.