Learn More
CPC Scientific 5TAMRA-Glu-Arg-Met-Arg-Pro-Arg-Lys-Arg-Gln-Gly-Ser-Val-Arg-Arg-Arg-Val-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110315
Encompass
ONE-LETTER SEQUENCE: 5TAMRA-ERMRPRKRQGSVRRRV-NH2MOLECULAR FORMULA: C108H176N42O24S1MOLECULAR WEIGHT:2478.9STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0315 Supplier CPC Scientific Supplier No. PROK051A
ONE-LETTER SEQUENCE: 5TAMRA-ERMRPRKRQGSVRRRV-NH2MOLECULAR FORMULA: C108H176N42O24S1MOLECULAR WEIGHT:2478.9STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases