Learn More
CPC Scientific 5TAMRA-His-Ala-Ala-Ile-Gly-Asp-Asp-Asp-Asp-Ala-Tyr-Ser-Ile-Thr-Ala-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110322
Encompass
ONE-LETTER SEQUENCE: 5TAMRA-HAAIGDDDDAYSITA-NH2MOLECULAR FORMULA: C89H116N20O30MOLECULAR WEIGHT:1946STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0322 Supplier CPC Scientific Supplier No. PROK054B
ONE-LETTER SEQUENCE: 5TAMRA-HAAIGDDDDAYSITA-NH2MOLECULAR FORMULA: C89H116N20O30MOLECULAR WEIGHT:1946STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases