Learn More
Cayman Chemical 5-trans Latanoprost (free acid), Purity: ≥95%, CAS: 903549-49-5, 5 mg
SDP
Catalog No. 502890409
Encompass_Preferred
An isomer of latanoprost (free acid) wherein the double bond between carbons 5 and 6 has been changed from cis (Z) to trans (E); intended primarily as an analytical standard
Catalog No. 50-289-0409 Supplier Cayman Chemical Supplier No. 101295 MG
An isomer of latanoprost (free acid) wherein the double bond between carbons 5 and 6 has been changed from cis (Z) to trans (E); intended primarily as an analytical standard