Learn More
Cayman Chemical 5-trans Fluprostenol isopropyl ester, Purity: ≥95%, CAS: 1563176-59-9, 50 mg
SDP
Catalog No. 502967161
Encompass_Preferred
A potential impurity in commercial preparations of travoprost
Catalog No. 50-296-7161 Supplier Cayman Chemical Supplier No. 1677750 MG
A potential impurity in commercial preparations of travoprost