Learn More
BIOCLONE INC 5µm BcMag™ Aldehyde-terminated magnetic beads | 150mg
Catalog No. NC1387998
Encompass_Preferred
BcMag™ Aldehyde-activated Magnetic Beads are uniform, silica-based superparamagnetic beads coated with high density aldehyde functional groups on the surface. The beads are used to covalently conjugate primary amine-containing ligands. BcMag™ Aldehyde-Terminated Magnetic Beads are most suitable for conjugation of large proteins. BcMagTM Long-arm Aldehyde-terminated Magnetic Beads are recommended for conjugation of small peptides because the long-arm (21-atom) hydrophilic linker may reduce steric hindrance.
Catalog No. NC1387998 Supplier BIOCLONE INC Supplier No. FE103
BcMag™ Aldehyde-activated Magnetic Beads are uniform, silica-based superparamagnetic beads coated with high density aldehyde functional groups on the surface. The beads are used to covalently conjugate primary amine-containing ligands. BcMag™ Aldehyde-Terminated Magnetic Beads are most suitable for conjugation of large proteins. BcMagTM Long-arm Aldehyde-terminated Magnetic Beads are recommended for conjugation of small peptides because the long-arm (21-atom) hydrophilic linker may reduce steric hindrance.