Learn More
Pluriselect Syringe-Strainer, 5 um mesh-size, 12 pcs. , sterile
Catalog No. NC1836208
Encompass_Preferred
The Syringe-strainer can be connected to a syringe or a tube via the screw cap with a Luer-Lock port. This product family comes in 15 different mesh sizes (1 um to 500 um) in a sterile or non-sterile variation. The syringe strainer can be used with positive or negative pressure.
Catalog No. NC1836208 Supplier Pluriselect Supplier No. 437100551
The Syringe-strainer can be connected to a syringe or a tube via the screw cap with a Luer-Lock port. This product family comes in 15 different mesh sizes (1 um to 500 um) in a sterile or non-sterile variation. The syringe strainer can be used with positive or negative pressure.