Learn More
Boston Bioproducts Inc RIPA Buffer (5X) - 100ml
Catalog No. NC9955727
Encompass
The 5X RIPA Buffer is used for rapid, efficient cell lysis and solubilization of proteins from adherent and suspension cultured mammalian cells and tissue samples. With the combined action of its ionic detergents, the RIPA buffer is extremely useful for disruption of all proteins within cellular compartments, including nuclear, membrane, and mitochondria. This buffer is compatible with many downstream applications, such as SDS-PAGE, immunoprecipitation, Western blot, and ELISA.
Catalog No. NC9955727 Supplier Boston Bioproducts Inc Supplier No. BP1155X100ML
The 5X RIPA Buffer is used for rapid, efficient cell lysis and solubilization of proteins from adherent and suspension cultured mammalian cells and tissue samples. With the combined action of its ionic detergents, the RIPA buffer is extremely useful for disruption of all proteins within cellular compartments, including nuclear, membrane, and mitochondria. This buffer is compatible with many downstream applications, such as SDS-PAGE, immunoprecipitation, Western blot, and ELISA.