Learn More
Cygnus Technologies Assay Diluent, 5X - Viral Clearance Prediction, Immuno-qPCR Application, 30 mL
Catalog No. NC2041299
Encompass
The Cygnus Technologies Assay Diluent, 5X, is a concentrated solution designed for the MockV™ MVM Kit Immuno-qPCR assay. It plays a vital role in viral clearance studies during biopharmaceutical manufacturing, utilizing non-infectious viral surrogates. This product stands out as an affordable and effective solution for assessing viral clearance across various stages of the biopharmaceutical life cycle.
Catalog No. NC2041299 Supplier Cygnus Technologies Supplier No. M00130
The Cygnus Technologies Assay Diluent, 5X, is a concentrated solution designed for the MockV™ MVM Kit Immuno-qPCR assay. It plays a vital role in viral clearance studies during biopharmaceutical manufacturing, utilizing non-infectious viral surrogates. This product stands out as an affordable and effective solution for assessing viral clearance across various stages of the biopharmaceutical life cycle.