Learn More
R&D Systems™ Cultrex 5X Collagen IV Solution
Description
Specifications
Specifications
| Content And Storage | Store in liquid nitrogen for up to 12 months from the date of receipt. |
| Quantity | 1 mL |
| Species | Multi-Species |
| Target | Collagen IV |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.