Learn More
Seracare Life Sciences Inc 5X DETECTOR BLOCK
Catalog No. 5067587
Encompass_Preferred
Detector Block is recommended as a general blocking agent for use in membrane-based detection assays. It is especially useful when traditional blocking solutions (I.e., milk, BSA, casein) reduce sensitivity or do not adequately block background. Detector Block can be used as both a blocking solution and a conjugate diluent in membrane applications such as Western, Northern, and Southern blotting.
This item is not returnable. View return policy
Catalog No. 50-675-87 Supplier Seracare Life Sciences Inc Supplier No. 59200004
Detector Block is recommended as a general blocking agent for use in membrane-based detection assays. It is especially useful when traditional blocking solutions (I.e., milk, BSA, casein) reduce sensitivity or do not adequately block background. Detector Block can be used as both a blocking solution and a conjugate diluent in membrane applications such as Western, Northern, and Southern blotting.