Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc DNA Gel Loading Solution (5X), Pack 5 X 10 mL
SDP

Catalog No. 50983253
Encompass_Preferred

A tracking dye during electrophoresis. Ensures the DNA samples remain submerged in the wells of the gel, by making them denser than the electrophoresis buffer.

Ready-to-use format
Two-color tracking of DNA migration
5X concentration
Filtered through a 0.22 µm membrane
Dnase, Rnase, and Protease tested

Use with agarose or non-denaturing polyacrylamide gel electrophoresis (PAGE) DNA gel electrophoresis Northern and Southern blot analysis Most commonly used to load and track DNA samples on electrophoretic gels.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Endotoxin and Mycoplasma Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50983253 Supplier Quality Biological Inc Supplier No. 351028661
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A tracking dye during electrophoresis. Ensures the DNA samples remain submerged in the wells of the gel, by making them denser than the electrophoresis buffer.

Ready-to-use format
Two-color tracking of DNA migration
5X concentration
Filtered through a 0.22 µm membrane
Dnase, Rnase, and Protease tested

Use with agarose or non-denaturing polyacrylamide gel electrophoresis (PAGE) DNA gel electrophoresis Northern and Southern blot analysis Most commonly used to load and track DNA samples on electrophoretic gels.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Endotoxin and Mycoplasma Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.