Learn More
Teknova 5X PBS-TBN Buffer. 1000mL, Sterile. 8 pack.
SDP
Catalog No. 502745850
Encompass
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.
Catalog No. 50-274-5850 Supplier Teknova Supplier No. P02118PK
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.