Learn More
Gold Biotechnology Inc 5X Phosphate Buffer, pH 8 250 mL
SDP
Catalog No. NC2107013
Encompass
Phosphate Buffer, pH8 is a mix of Sodium Phosphate dibasic (Na2HPO4) and Monosodium Phosphate (NaH2PO4). This buffer is the basis for wash and elution buffers for protein purification through His-tag.
This item is not returnable. View return policy
Catalog No. NC2107013 Supplier Gold Biotechnology Inc Supplier No. P500250
Phosphate Buffer, pH8 is a mix of Sodium Phosphate dibasic (Na2HPO4) and Monosodium Phosphate (NaH2PO4). This buffer is the basis for wash and elution buffers for protein purification through His-tag.