Learn More
National Diagnostics 5X Protein Loading Buffer, 10x1mL
SDP
Catalog No. NC1621853
Encompass
5X Protein Loading Buffer is a reducing sample buffer for SDS-polyacrylamide gel electrophoresis. 5X Protein Loading Buffer contains 1.0M TrisHCl (pH 8.5), 8% (w/v) lithium dodecyl sulfate, 40% (v/w) glycerol, 2mM EDTA, 0.5M DTT and tracking dye in distilled/deionized water.
Catalog No. NC1621853 Supplier National Diagnostics Supplier No. EC887
5X Protein Loading Buffer is a reducing sample buffer for SDS-polyacrylamide gel electrophoresis. 5X Protein Loading Buffer contains 1.0M TrisHCl (pH 8.5), 8% (w/v) lithium dodecyl sulfate, 40% (v/w) glycerol, 2mM EDTA, 0.5M DTT and tracking dye in distilled/deionized water.