Learn More
Kuhner Shaker Inc 5X REPL SOLID SILICONE SEAL
Catalog No. NC2399183
Encompass
5x Replacement solid silicone seal, 1.2 mm LOW EVAP holes, for universal cover 24-well MTPs, 18 mm hole distance (CR1324b)
Catalog No. NC2399183 Supplier Kuhner Shaker Inc Supplier No. SMCR0008A
5x Replacement solid silicone seal, 1.2 mm LOW EVAP holes, for universal cover 24-well MTPs, 18 mm hole distance (CR1324b)