Learn More
Kuhner Shaker Inc 5X REPLACEMENT SPONGY SILICONE
Supplier:  Kuhner Shaker Inc SMCR0007
Encompass
5x Replacement spongy silicone seal, 2.5mm holes for sandwich cover for 24 deepwell MTP (CR1224, CR1224b)
Catalog No. NC1450759