Learn More
Cell Biolabs Inc 5X RIPA Buffer, 20 mL, with Protease Inhibitor Cocktail
Supplier:  Cell Biolabs Inc AKR190
Encompass_Preferred
RIPA Buffer continues to be a popular buffer to lyse plated and suspension cultured mammalian cells. The buffer extracts cytoplasmic, membrane, and nuclear proteins and is suitable for downstream assays such as reporter assays, protein assays, immunoassays and protein purification. 20 mL containing 125 mM Tris pH 7.6, 750 mM NaCl, 5% NP-40, 5% sodium deoxycholate, 0.5% SDS. A separate protease inhibitor cocktail is included.
Catalog No. 50-672-584