Learn More
Genscript Corporation 5X Sample Buffer
Catalog No. NC0879818
Encompass
5X Sample Buffer is used as a tracking dye for SDS PAGE gel loading. This buffer contains SDS and is suitable for denaturing gel electrophoresis. Dilute 1:4 with sample before loading.
Catalog No. NC0879818 Supplier Genscript Corporation Supplier No. MB01015
5X Sample Buffer is used as a tracking dye for SDS PAGE gel loading. This buffer contains SDS and is suitable for denaturing gel electrophoresis. Dilute 1:4 with sample before loading.