Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc SDS Protein Gel Loading Solution (5X), 5 mL
SDP

Catalog No. 50447713
Encompass_Preferred

Formulated as 5X rather than the traditional 2X stocks, SDS Protein Gel Loading Solution (5X) enables a larger volume of sample to be loaded per well.

5X concentrated protein gel loading buffer

Applications: Simply load the gel sample wells and visualize progress of the electrophoresis run by monitoring the location of the blue dye front. Lane Marker Sample Buffers may be used in denaturing gels of all kinds and are compatible with coomassie dye and silver staining techniques, as well as Western blotting procedures. SDS Protein Gel Loading Solution (5X) is useful for SDS-PAGE and Western Blotting.

Storage Conditions: 15 °C to 30 °C
Shipping Conditions: ambient temperature
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content Product and/or packaging is recyclable or reduces waste Product and/or packaging is safer to human and environmental health

Catalog No. 50-447-713 Supplier Quality Biological Inc Supplier No. 351334021EA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formulated as 5X rather than the traditional 2X stocks, SDS Protein Gel Loading Solution (5X) enables a larger volume of sample to be loaded per well.

5X concentrated protein gel loading buffer

Applications: Simply load the gel sample wells and visualize progress of the electrophoresis run by monitoring the location of the blue dye front. Lane Marker Sample Buffers may be used in denaturing gels of all kinds and are compatible with coomassie dye and silver staining techniques, as well as Western blotting procedures. SDS Protein Gel Loading Solution (5X) is useful for SDS-PAGE and Western Blotting.

Storage Conditions: 15 °C to 30 °C
Shipping Conditions: ambient temperature
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content Product and/or packaging is recyclable or reduces waste Product and/or packaging is safer to human and environmental health

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.