Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED 5X siRNA Buffer Solution, 100ml
Catalog No. NC1338268
Encompass
Concentrated buffer solution recommended for resuspension and long-term storage of any short, double-strand, or single-strand synthetic RNA molecule. Dilute with RNase-free water prior to use.
This item is not returnable. View return policy
Catalog No. NC1338268 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. B002000UB100
Concentrated buffer solution recommended for resuspension and long-term storage of any short, double-strand, or single-strand synthetic RNA molecule. Dilute with RNase-free water prior to use.