Learn More
Teknova 5X TBE DNA Sequencing Grade Solution. 4L, Non-Sterile. 4 pack.
SDP
Catalog No. 502387491
Encompass
445mM Tris, 445mM Boric Acid, 10mM Ethylenediaminetetraacetic acid (EDTA)
Catalog No. 50-238-7491 Supplier Teknova Supplier No. T02754PK
445mM Tris, 445mM Boric Acid, 10mM Ethylenediaminetetraacetic acid (EDTA)