Learn More
Immunochemistry Technology 5X UNIVERSAL PLATE COATING BUF
Catalog No. NC9170781
Encompass
CB1, CB1 is a unique buffer used to coat antibodies and antigens onto polystyrene microtiter ELISA plates, For use in antibody-sandwich ELISAs and antigen-down ELISAs, 100mL
Catalog No. NC9170781 Supplier Immunochemistry Technology Supplier No. 644
CB1, CB1 is a unique buffer used to coat antibodies and antigens onto polystyrene microtiter ELISA plates, For use in antibody-sandwich ELISAs and antigen-down ELISAs, 100mL