Learn More
Ace Glass, Inc. 5mm OD x 135mm porosity E filter stick
SDP
Catalog No. 50976772
Encompass_Preferred
5mm O.D. x 135mm gas dispersion tube for fine dispersion. Porous tip is approximately 5mm O.D. x 10mm. Supplied with porosity E (4-8 micron) filter stick.
Catalog No. 50-976-772 Supplier Ace Glass, Inc. Supplier No. 943510
5mm O.D. x 135mm gas dispersion tube for fine dispersion. Porous tip is approximately 5mm O.D. x 10mm. Supplied with porosity E (4-8 micron) filter stick.