Learn More
Takara Bio HisTALON™ Gravity Columns, 5 x 1 mL Columns
SDP
Catalog No. NC1612157
Encompass
Prepacked gravity columns, each containing 1 ml of TALON Resin, an immobilized metal affinity chromatography resin for the purification of recombinant his-tagged proteins. These columns are suitable for gravity flow purification under native or denaturing conditions.
Catalog No. NC1612157 Supplier Takara Bio Supplier No. 635655
Prepacked gravity columns, each containing 1 ml of TALON Resin, an immobilized metal affinity chromatography resin for the purification of recombinant his-tagged proteins. These columns are suitable for gravity flow purification under native or denaturing conditions.