Learn More
Ace Glass, Inc. 5mm OD x 210mm porosity D filter stick
SDP
Catalog No. 50979998
Encompass_Preferred
5mm O.D. x 210mm gas dispersion tube for fine dispersion. Porous tip is approximately 5mm O.D. x 10mm. Supplied with porosity D (10-20 micron) filter stick.
Catalog No. 50-979-998 Supplier Ace Glass, Inc. Supplier No. 943539
5mm O.D. x 210mm gas dispersion tube for fine dispersion. Porous tip is approximately 5mm O.D. x 10mm. Supplied with porosity D (10-20 micron) filter stick.