Learn More
GNOSKO BIO INC 5x5 Clear Polymer Freezer Rack
Catalog No. 503126229
Encompass_Preferred
ChillView freezer racks address common issues with stainless steel freezer boxes. ChillView racks are clear and don't frost, so they don't require gloves to handle. Everything can be seen when opening the door, greatly reducing the time and energy wasted when searching for something specific. Energy is also greatly reduced since ChillView racks transfer heat at approx. 100x less than the rate of stainless-steel boxes. ChillView also allows RFID tags to be read through the box to enable Gnosko Bio automated inventory management solutions. 5 shelves, 5 boxes deep.
Catalog No. 50-312-6229 Supplier GNOSKO BIO INC Supplier No. GB00CV3
Add to Cart
Add to Cart
ChillView freezer racks address common issues with stainless steel freezer boxes. ChillView racks are clear and don't frost, so they don't require gloves to handle. Everything can be seen when opening the door, greatly reducing the time and energy wasted when searching for something specific. Energy is also greatly reduced since ChillView racks transfer heat at approx. 100x less than the rate of stainless-steel boxes. ChillView also allows RFID tags to be read through the box to enable Gnosko Bio automated inventory management solutions. 5 shelves, 5 boxes deep.