Learn More
USA Scientific Inc 25-place (5 x 5) upright freezer sliding rack for 2inch H boxes
Catalog No. NC1148521
Encompass
25-place (5 long x 5 high) upright freezer sliding rack for 2" H boxes, 26 3/4" L x 5 1/2" W x 11 4/5" H, stainless steel
Catalog No. NC1148521 Supplier USA Scientific Inc Supplier No. 26325500
25-place (5 long x 5 high) upright freezer sliding rack for 2" H boxes, 26 3/4" L x 5 1/2" W x 11 4/5" H, stainless steel