Learn More
Sigma Aldrich Fine Chemicals Biosciences Dexamethasone, 50-02-2, MFCD00064136, 25 mg
Catalog No. NC1519413
Encompass
Empirical Formula (Hill Notation): C22H29FO5, Molecular Weight: 392.46, powder, BioReagent, suitable for cell culture, ≥97%, Synonym: (11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F. Dexamethasone is used to study apoptosis, cell signaling pathways, cell differentiation and gene expression.
Catalog No. NC1519413 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D490225MG
Empirical Formula (Hill Notation): C22H29FO5, Molecular Weight: 392.46, powder, BioReagent, suitable for cell culture, ≥97%, Synonym: (11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F. Dexamethasone is used to study apoptosis, cell signaling pathways, cell differentiation and gene expression.