Learn More
Andwin Scientific 5X7 BLUE BASIC AUTORAD FILM
SDP
Catalog No. NC9167068
Encompass_Preferred
The 5" x 7" GeneMate blue autoradiography film is ideal for different applications like the chemiluminescence and autoradiography of 32P and 125I. This product lasts 2 years from the date of the manufacturing. It is sold in a box of 100.
Catalog No. NC9167068 Supplier Andwin Scientific Supplier No. F90235X7
The 5" x 7" GeneMate blue autoradiography film is ideal for different applications like the chemiluminescence and autoradiography of 32P and 125I. This product lasts 2 years from the date of the manufacturing. It is sold in a box of 100.