Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies 5x 930 dsDNA Inlet Buffer, Molecular Biology, 300 mL

Catalog No. NC2943887
Encompass

The 5x 930 dsDNA Inlet Buffer is a concentrated running buffer formulated for use with Agilent Fragment Analyzer systems, specifically optimized for the analysis of dsDNA fragments between 75 bp and 20 kb. This reagent facilitates accurate sizing and relative quantification of samples during electrophoresis. It is intended for research use only and must not be used in diagnostic procedures.

  • Concentration: 5x
  • Volume: 300 mL
  • Application: Nucleic acid quantification and molecular biology research
  • Storage: Store at 4°C; do not freeze
  • pH range: 7.15 to 7.55

Catalog No. NC2943887 Supplier Agilent Technologies Supplier No. DNF3550300
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 5x 930 dsDNA Inlet Buffer is a concentrated running buffer formulated for use with Agilent Fragment Analyzer systems, specifically optimized for the analysis of dsDNA fragments between 75 bp and 20 kb. This reagent facilitates accurate sizing and relative quantification of samples during electrophoresis. It is intended for research use only and must not be used in diagnostic procedures.

  • Concentration: 5x
  • Volume: 300 mL
  • Application: Nucleic acid quantification and molecular biology research
  • Storage: Store at 4°C; do not freeze
  • pH range: 7.15 to 7.55

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.