Learn More
Gold Biotechnology Inc 5X Phosphate Buffer + 3M Imidazole Solution 100 mL
SDP
Catalog No. NC2107017
Encompass
This Buffer set allows researchers to develop, wash and elute buffers for His-tag protein purification.
This item is not returnable. View return policy
Catalog No. NC2107017 Supplier Gold Biotechnology Inc Supplier No. I905100
This Buffer set allows researchers to develop, wash and elute buffers for His-tag protein purification.