Learn More
Cayman Chemical 5Z 11Z 15R15HydroxyeIcosa
SDP
Catalog No. 502984241
Encompass_Preferred
A stable isomer of 15(S)-HETE that elicits contraction of isolated pulmonary arteries from rabbits and inhibits the proliferation and migration of PC-3 cells
Catalog No. 50-298-4241 Supplier Cayman Chemical Supplier No. 19557500 UG
A stable isomer of 15(S)-HETE that elicits contraction of isolated pulmonary arteries from rabbits and inhibits the proliferation and migration of PC-3 cells