Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (5Z,2E)-CU-3 | 1815598-71-0 | 95.0% | 1 ML
SDP

Catalog No. 5000232454
Encompass_Preferred

(5Z,2E)-CU-3 is a small-molecule diacylglycerol kinase (DGK) inhibitor and apoptosis inducer supplied as a ready-to-use 10 mM solution in DMSO for research applications.

  • Supplied as a 10 mM solution in DMSO (1 mL).
  • High purity verified at 95.0%.
  • Molecular weight 392.47 g/mol and formula C16H12N2O4S3.
  • Solid, yellow to brown appearance.
  • Intended for apoptosis and DGK pathway research.

Catalog No. 50-002-32454 Supplier Medchemexpress LLC Supplier No. HY121638A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(5Z,2E)-CU-3 is a small-molecule diacylglycerol kinase (DGK) inhibitor and apoptosis inducer supplied as a ready-to-use 10 mM solution in DMSO for research applications.

  • Supplied as a 10 mM solution in DMSO (1 mL).
  • High purity verified at 95.0%.
  • Molecular weight 392.47 g/mol and formula C16H12N2O4S3.
  • Solid, yellow to brown appearance.
  • Intended for apoptosis and DGK pathway research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.