Learn More
Cayman Chemical (5Z,2E)-CU-3, Purity: ≥98%, CAS: 1815598-71-0, 25 mg
SDP
Catalog No. 502916704
Encompass_Preferred
A DGK-α inhibitor (IC50 = 0.6 µM for the porcine enzyme); selective for DGK-α over other type I DGKs, as well as type II, type III, type IV, and type V DGKs (IC50s = 7-36 µM); induces apoptosis in HeLa and HepG2 cells and enhances concanavalin A-induced increases in IL2 mRNA expression in Jurkat T cells at 5 µM
Catalog No. 50-291-6704 Supplier Cayman Chemical Supplier No. 3777025 MG
A DGK-α inhibitor (IC50 = 0.6 µM for the porcine enzyme); selective for DGK-α over other type I DGKs, as well as type II, type III, type IV, and type V DGKs (IC50s = 7-36 µM); induces apoptosis in HeLa and HepG2 cells and enhances concanavalin A-induced increases in IL2 mRNA expression in Jurkat T cells at 5 µM