Learn More
Medchemexpress LLC (5Z)-5-[(3,5-difluoro-4-hydroxyphenyl)methylene]-3,5-dihydro-2-methyl-3-(2,2,2-trifluoroethyl)-4h-imid | 1539318-36-9 | MFCD29081187 | 98.1% | 320.21 g/mol | C13H9F5N2O2 | 5 MG
SDP
Catalog No. 5000263676
Encompass_Preferred
DFHBI-1T is a membrane-permeable RNA-aptamer-activated fluorescent probe that produces bright, low-background fluorescence (ex/em = 472 nm/507 nm) when bound to target RNA aptamers. Supplied as a light-yellow solid with high purity, it is suitable for live-cell RNA imaging and in vitro fluorescence assays.
  • Membrane-permeable fluorophore suited for live-cell imaging
  • Activates fluorescence upon binding specific RNA aptamers, reducing background
  • Excitation/emission maxima approximately 472 nm/507 nm
  • High purity for reproducible experimental results
  • Solid form with convenient small-scale packaging for assay development
  • Compatible with common fluorescence filter sets
Catalog No. 50-002-63676 Supplier Medchemexpress LLC Supplier No. HY1102515MG
DFHBI-1T is a membrane-permeable RNA-aptamer-activated fluorescent probe that produces bright, low-background fluorescence (ex/em = 472 nm/507 nm) when bound to target RNA aptamers. Supplied as a light-yellow solid with high purity, it is suitable for live-cell RNA imaging and in vitro fluorescence assays.
  • Membrane-permeable fluorophore suited for live-cell imaging
  • Activates fluorescence upon binding specific RNA aptamers, reducing background
  • Excitation/emission maxima approximately 472 nm/507 nm
  • High purity for reproducible experimental results
  • Solid form with convenient small-scale packaging for assay development
  • Compatible with common fluorescence filter sets