Learn More
Medchemexpress LLC 5Z-7-Oxozeaenol | 253863-19-3 | 99.3% | 1 ML
SDP
Catalog No. 5000338700
Encompass_Preferred
5Z-7-Oxozeaenol is a natural anti-protozoan compound originating from fungi. It functions as a potent, irreversible, and selective inhibitor of TAK1 and VEGF-R2, which helps prevent inflammation. This compound is intended for research applications only.
  • Potent, irreversible, and selective inhibitor of TAK1 and VEGF-R2
  • Inhibits the JNK/p38 pathway in a signal-specific manner
  • Suppresses PMA-induced AP-1 activity in KT cells
  • Shows inhibitory activity against VEGF-R3, FLT3, PDGFR-β, B-RAF VE, and SRC
Catalog No. 50-003-38700 Supplier Medchemexpress LLC Supplier No. HY1268610MM/1ML
5Z-7-Oxozeaenol is a natural anti-protozoan compound originating from fungi. It functions as a potent, irreversible, and selective inhibitor of TAK1 and VEGF-R2, which helps prevent inflammation. This compound is intended for research applications only.
  • Potent, irreversible, and selective inhibitor of TAK1 and VEGF-R2
  • Inhibits the JNK/p38 pathway in a signal-specific manner
  • Suppresses PMA-induced AP-1 activity in KT cells
  • Shows inhibitory activity against VEGF-R3, FLT3, PDGFR-β, B-RAF VE, and SRC