Learn More
Medchemexpress LLC 5Z-7-Oxozeaenol | 253863-19-3 | 99.3% | 25 MG
SDP
Catalog No. 5000338709
Encompass_Preferred
5Z-7-Oxozeaenol is a natural anti-protozoan compound of fungal origin. It acts as a potent, irreversible, and selective inhibitor of TAK1 and VEGF-R2, with IC50s of 8 nM and 52 nM, respectively. This product is for research use only.
  • Potent and selective inhibitor of TAK1 (IC50 8.1 nM).
  • Inhibits VEGF-R2, VEGF-R3, FLT3, PDGFR-β, B-RAF VE, and SRC.
  • Prevents inflammation by inhibiting the catalytic activity of TAK1 MAPK kinase kinase.
  • Inhibits JNK/p38 pathway in a signal-specific manner.
  • Exhibits antiproliferative and cytotoxic activities against various cell lines.
Catalog No. 50-003-38709 Supplier Medchemexpress LLC Supplier No. 25MGHY12686
5Z-7-Oxozeaenol is a natural anti-protozoan compound of fungal origin. It acts as a potent, irreversible, and selective inhibitor of TAK1 and VEGF-R2, with IC50s of 8 nM and 52 nM, respectively. This product is for research use only.
  • Potent and selective inhibitor of TAK1 (IC50 8.1 nM).
  • Inhibits VEGF-R2, VEGF-R3, FLT3, PDGFR-β, B-RAF VE, and SRC.
  • Prevents inflammation by inhibiting the catalytic activity of TAK1 MAPK kinase kinase.
  • Inhibits JNK/p38 pathway in a signal-specific manner.
  • Exhibits antiproliferative and cytotoxic activities against various cell lines.