Learn More
Medchemexpress LLC 5Z,8Z,11Z,14Z-Eicosatetraenoic-16,16,17,17,18,18,19,19,20,20,20-d11 acid | 2692624-11-4 | 98.0% | 315.53 | 1 ML
SDP
Catalog No. 5000345005
Encompass_Preferred
Arachidonic acid-d11 is the deuterium labeled Arachidonic acid. Arachidonic acid is an essential fatty acid and a major constituent of biomembranes. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Incorporated into drug molecules as tracers for quantitation during drug development.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-45005 Supplier Medchemexpress LLC Supplier No. HY109590S2100UG
Arachidonic acid-d11 is the deuterium labeled Arachidonic acid. Arachidonic acid is an essential fatty acid and a major constituent of biomembranes. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Incorporated into drug molecules as tracers for quantitation during drug development.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs.