Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5Z,8Z,11Z,14Z-Eicosatetraenoic-16,16,17,17,18,18,19,19,20,20,20-d11 acid | 2692624-11-4 | 98.0% | 315.53 | 100 MG
SDP

Catalog No. 5000345140
Encompass_Preferred

Arachidonic acid-d11 is the deuterium labeled Arachidonic acid, an essential fatty acid and a major constituent of biomembranes. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs. This product has been tested and complies with the given specifications. Solution, -20°C, 2 years.

  • Target: Endogenous Metabolite; Isotope-Labeled Compounds
  • Pathway: Metabolic Enzyme/Protease; Others
  • Appearance: Colorless to light yellow (Liquid)

Catalog No. 50-003-45140 Supplier Medchemexpress LLC Supplier No. HY109590S2500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arachidonic acid-d11 is the deuterium labeled Arachidonic acid, an essential fatty acid and a major constituent of biomembranes. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs. This product has been tested and complies with the given specifications. Solution, -20°C, 2 years.

  • Target: Endogenous Metabolite; Isotope-Labeled Compounds
  • Pathway: Metabolic Enzyme/Protease; Others
  • Appearance: Colorless to light yellow (Liquid)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.