Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5Z,8Z,11Z,14Z-Eicosatetraenoic-16,16,17,17,18,18,19,19,20,20,20-d11 acid | 2692624-11-4 | 98.0% | 315.53 | 50 MG
SDP

Catalog No. 5000345139
Encompass_Preferred

Arachidonic acid-d11 is the deuterium labeled Arachidonic acid. It is an essential fatty acid and a major constituent of biomembranes. This product is provided as a colorless to light yellow liquid solution.

  • Deuterium labeled
  • Essential fatty acid
  • Major constituent of biomembranes
  • Colorless to light yellow liquid
  • Purity of 98.0%
  • Store at -20°C for up to 2 years

Catalog No. 50-003-45139 Supplier Medchemexpress LLC Supplier No. HY109590S21MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arachidonic acid-d11 is the deuterium labeled Arachidonic acid. It is an essential fatty acid and a major constituent of biomembranes. This product is provided as a colorless to light yellow liquid solution.

  • Deuterium labeled
  • Essential fatty acid
  • Major constituent of biomembranes
  • Colorless to light yellow liquid
  • Purity of 98.0%
  • Store at -20°C for up to 2 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.