Learn More
Medchemexpress LLC (5Z,8Z,14Z)-11,12-Dihydroxyicosa-5,8,14-trienoic acid-d11 | 99.9% | 349.55 | 25 UG
SDP
Catalog No. 5000439444
Encompass_Preferred
This product is a deuterated form of 11,12-DiHETrE, a dihydroxy metabolite of arachidonic acid. It features eleven deuterium atoms, making it suitable for use as an internal standard in mass spectrometry-based quantitative analysis. This high-purity compound is provided as a solution for research applications.
  • High purity
  • Isotopically labeled
  • Suitable for mass spectrometry
  • Stable in solution
  • Research use only
Catalog No. 50-004-39444 Supplier Medchemexpress LLC Supplier No. 25UGHY113111S
This product is a deuterated form of 11,12-DiHETrE, a dihydroxy metabolite of arachidonic acid. It features eleven deuterium atoms, making it suitable for use as an internal standard in mass spectrometry-based quantitative analysis. This high-purity compound is provided as a solution for research applications.
  • High purity
  • Isotopically labeled
  • Suitable for mass spectrometry
  • Stable in solution
  • Research use only