Learn More
Cayman Chemical 5Z8Z11ZHeptadecatrIe250ug
SDP
Catalog No. 502914744
Encompass_Preferred
An internal standard for the quantification of 5(Z),8(Z),11(Z)-heptadecatrienoic acid by GC- or LC-MS
Catalog No. 50-291-4744 Supplier Cayman Chemical Supplier No. 39690250 UG
An internal standard for the quantification of 5(Z),8(Z),11(Z)-heptadecatrienoic acid by GC- or LC-MS